MilliporeSigma
  • Highly enantioselective asymmetric hydrogenation of alpha-phthalimide ketone: an efficient entry to enantiomerically pure amino alcohols.
Highly enantioselective asymmetric hydrogenation of alpha-phthalimide ketone: an efficient entry to enantiomerically pure amino alcohols.
Journal of the American Chemical Society (2004-02-12)
Aiwen Lei, Shulin Wu, Minsheng He, Xumu Zhang
ABSTRACT
A new type of alpha-phthalimide ketones was hydrogenated in excellent enantioselectivity by using a Ru-(C3-TunePhos) complex as the catalyst. Up to 10 000 turnovers have been achieved in more than 99% ee in the hydrogenation reaction. A dynamic kinetic resolution study for the synthesis of threonine was performed, and high anti selectivity (>97:3) was observed for the first time. An efficient method to synthesize enantiomerically pure amino alcohols has been developed.