MilliporeSigma
  • Palladium-catalyzed cross-coupling reactions of heterocyclic silanolates with substituted aryl iodides and bromides.
Palladium-catalyzed cross-coupling reactions of heterocyclic silanolates with substituted aryl iodides and bromides.
Organic letters (2006-02-14)
Scott E Denmark, John D Baird
ABSTRACT
[reaction: see text] Sodium silanolates derived from a number of heterocyclic silanols undergo cross-coupling with a variety of aromatic iodides and bromides under mild conditions. In situ deprotonation of the silanols with an equivalent amount of sodium hydride in toluene generates the sodium salt that couples with iodides under the action of Pd(2)(dba)(3).CHCl(3) in good yield at room temperature to 50 degrees C. The aromatic bromides also couple with these salts under the action of the Pd(I) catalyst 12.