MilliporeSigma
  • Impact of collection, isolation and storage methodology of circulating microvesicles on flow cytometric analysis.
Impact of collection, isolation and storage methodology of circulating microvesicles on flow cytometric analysis.
Experimental and therapeutic medicine (2015-12-17)
Fancong Kong, Liming Zhang, Hongxiang Wang, Guolin Yuan, Anyuan Guo, Qiubai Li, Zhichao Chen
ABSTRACT
Microvesicles (MVs) in body fluids participate in a variety of physical and pathological processes, and are regarded as potential biomarkers for numerous diseases. Flow cytometry (FCM) is among the most frequently used techniques for MV detection. However, different handling methods unavoidably cause pre-analytical variations in the counts and sizes of MVs determined by FCM. The aim of the present study was to investigate the effect of centrifugation, storage conditions and anticoagulant on MV measurements. Blood samples were obtained from 13 healthy donors, including 4 women and 9 men. Calcein-AM staining was used to label MVs and assess the impact of pre-analytical preparation, including centrifugation, and storage conditions on MV measurements obtained using FCM. The range of factors investigated for comparison included: Platelet-free plasma (PFP) stored at -80°C for 1 or 4 weeks; MVs stored at 4°C for 3-4 days or 1 week; MVs frozen at -80°C for 1 or 4 weeks; and anticoagulants, either heparin or ethylenediaminetetraacetic acid (EDTA). No statistically significant differences in MV counts were detected between the two centrifugation speeds (16,000 and 20,500 × g) or among the three centrifugation times (15, 30 and 60 min) investigated. Similarly, no significant differences were noted in MV counts between the two anticoagulants tested (heparin and EDTA). However, the storage of PFP or MVs in heparin-anticoagulated plasma for different periods markedly affected the detected MV counts and size distribution. The counts and sizes of MVs from EDTA-anticoagulated plasma were only affected when the MVs were frozen at -80°C for 4 weeks. In conclusion, calcein-AM is able to efficiently identify MVs from plasma and may be an alternative to Annexin V for MV staining. EDTA preserves the MV counts and size more accurately compared with heparin under calcein-AM staining. PFP centrifuged at 16,000 × g for 15 min is sufficient to isolate MVs, which enables the batch processing of samples. PFP, rather than MVs alone, appears to be the preferable mode of sample storage, as MVs stored in PFP were less affected by storage temperature and duration. The present study provides a methodology for MV collection, storage and isolation, to facilitate further investigation of MVs as biomarkers in disease.
Sigma-Aldrich
Latex beads, amine-modified polystyrene, fluorescent yellow-green, aqueous suspension, 1.0 μm mean particle size
Latex beads, polystyrene, 3.0 μm mean particle size