MilliporeSigma
  • Production of the novel two-peptide lantibiotic lichenicidin by Bacillus licheniformis DSM 13.
Production of the novel two-peptide lantibiotic lichenicidin by Bacillus licheniformis DSM 13.
PloS one (2009-08-27)
Jasmin Dischinger, Michaele Josten, Christiane Szekat, Hans-Georg Sahl, Gabriele Bierbaum
ABSTRACT
Lantibiotics are small microbial peptide antibiotics that are characterized by the presence of the thioether amino acids lanthionine and methyllanthionine. Lantibiotics possess structural genes which encode inactive prepeptides. During maturation, the prepeptide undergoes posttranslational modifications including the introduction of rare amino acids as lanthionine and methyllanthione as well as the proteolytic removal of the leader. The structural gene (lanA) as well as the other genes which are involved in lantibiotic modification (lanM, lanB, lanC, lanP), regulation (lanR, lanK), export (lanT(P)) and immunity (lanEFG) are organized in biosynthetic gene clusters. Sequence comparisons in the NCBI database showed that Bacillus licheniformis DSM 13 harbours a putative lantibiotic gene cluster which comprises two structural genes (licA1, licA2) and two modification enzymes (licM1, licM2) in addition to 10 ORFs that show sequence similarities to proteins involved in lantibiotic production. A heat labile antimicrobial activity was detected in the culture supernatant and a heat stabile activity was present in the isopropanol cell wash extract of this strain. In agar well diffusion assays both fractions exhibited slightly different activity spectra against Gram-positive bacteria. In order to demonstrate the connection between the lantibiotic gene cluster and one of the antibacterial activities, two Bacillus licheniformis DSM 13 mutant strains harbouring insertions in the structural genes of the modification enzymes licM1 and licM2 were constructed. These strains were characterized by a loss of activity in the isopropanol extract and substractive MALDI-TOF predicted masses of 3020.6 Da and 3250.6 Da for the active peptides. In conclusion, B. licheniformis DSM 13 produces an antimicrobial substance that represents the two-peptide lantibiotic lichenicidin and that shows activity against a wide range of Gram-positive bacteria including methicillin resistant Staphylococcus aureus strains.
Sigma-Aldrich
4-Hydroxy-L-isoleucine, ≥98.0% (TLC)