MilliporeSigma
  • Gas-phase fragmentation of oligoproline peptide ions lacking easily mobilizable protons.
Gas-phase fragmentation of oligoproline peptide ions lacking easily mobilizable protons.
Journal of the American Society for Mass Spectrometry (2013-04-24)
Magdalena Rudowska, Robert Wieczorek, Alicja Kluczyk, Piotr Stefanowicz, Zbigniew Szewczuk
ABSTRACT
The fragmentation of peptides containing quaternary ammonium group, but lacking easily mobilizable protons, was examined with the aid of deuterium-labeled analogs and quantum-chemical modeling. The fragmentation of oligoproline containing quaternary ammonium group involves the mobilization of hydrogens localized at α- and γ- or δ-carbon atoms in the pyrrolidine ring of proline. The study of the dissociation pattern highlights the unusual proline residue behavior during MS/MS experiments of peptides.
Sigma-Aldrich
Fmoc-Pro-OH, ≥90% (HPLC)