MilliporeSigma
  • Cross-species functional genomic analysis identifies resistance genes of the histone deacetylase inhibitor valproic acid.
Cross-species functional genomic analysis identifies resistance genes of the histone deacetylase inhibitor valproic acid.
PloS one (2012-11-17)
Rakel Brendsdal Forthun, Tanima Sengupta, Hanne Kim Skjeldam, Jessica Margareta Lindvall, Emmet McCormack, Bjørn Tore Gjertsen, Hilde Nilsen
ABSTRACT
The mechanisms of successful epigenetic reprogramming in cancer are not well characterized as they involve coordinated removal of repressive marks and deposition of activating marks by a large number of histone and DNA modification enzymes. Here, we have used a cross-species functional genomic approach to identify conserved genetic interactions to improve therapeutic effect of the histone deacetylase inhibitor (HDACi) valproic acid, which increases survival in more than 20% of patients with advanced acute myeloid leukemia (AML). Using a bidirectional synthetic lethality screen revealing genes that increased or decreased VPA sensitivity in C. elegans, we identified novel conserved sensitizers and synthetic lethal interactors of VPA. One sensitizer identified as a conserved determinant of therapeutic success of HDACi was UTX (KDM6A), which demonstrates a functional relationship between protein acetylation and lysine-specific methylation. The synthetic lethal screen identified resistance programs that compensated for the HDACi-induced global hyper-acetylation, and confirmed MAPKAPK2, HSP90AA1, HSP90AB1 and ACTB as conserved hubs in a resistance program for HDACi that are drugable in human AML cell lines. Hence, these resistance hubs represent promising novel targets for refinement of combinatorial epigenetic anti-cancer therapy.
Sigma-Aldrich
MISSION® esiRNA, targeting human KDM6A (1)
Pricing and availability is not currently available.