MilliporeSigma
  • Transfection of poly(I:C) can induce reactive oxygen species-triggered apoptosis and interferon-β-mediated growth arrest in human renal cell carcinoma cells via innate adjuvant receptors and the 2-5A system.
Transfection of poly(I:C) can induce reactive oxygen species-triggered apoptosis and interferon-β-mediated growth arrest in human renal cell carcinoma cells via innate adjuvant receptors and the 2-5A system.
Molecular cancer (2014-09-18)
Nanae Harashima, Takafumi Minami, Hirotsugu Uemura, Mamoru Harada
ABSTRACT
Synthetic double-stranded RNA poly(I:C) is a useful immune adjuvant and exhibits direct antitumor effects against several types of cancers. In this study, we elucidated the mechanisms underlying the effects induced in poly(I:C)-transfected human renal cell carcinoma (RCC) cells. In contrast to the lack of an effect of adding poly(I:C), poly(I:C) transfection drastically decreased RCC cell viability. Poly(I:C) transfection induced reactive oxygen species (ROS)-dependent apoptosis in RCC cells and decreased the mitochondrial membrane potential (ΔΨm). Treatment with N-acetyl-L-cysteine (NAC), a ROS scavenger, suppressed apoptosis and restored the ΔΨm. Although the levels of phosphorylated γH2A.X, an indicator of DNA damage, increased in poly(I:C)-transfected RCC cells, NAC treatment decreased their levels, suggesting ROS-mediated DNA damage. Furthermore, poly(I:C) transfection increased the levels of phosphorylated p53, NOXA, and tBid. Immunoblots and assays with a panel of caspase inhibitors revealed that poly(I:C) transfection-induced apoptosis was dependent on caspase-8 and -9, as well as caspase-2. Alternatively, poly(I:C) transfection increased mRNA expression of interferon (IFN)-β, and treatment with IFN-β suppressed growth of RCC cells without apoptosis. In addition, cyclinD1 and c-Myc expression decreased in poly(I:C)-transfected RCC cells. Moreover, RNA interference experiments revealed that poly(I:C) transfection exerted apoptotic effects on RCC cells through innate adjuvant receptors and the 2-5A system, the latter of which induces apoptosis in virus-infected cells. These results suggest that poly(I:C) transfection induced two types of effects against RCC cells such as apoptosis, as a result of ROS-mediated DNA damage, and IFN-β-mediated growth arrest, both of which were exerted via innate adjuvant receptors and the 2-5A system.
Sigma-Aldrich
MISSION® esiRNA, targeting human IRF3