MilliporeSigma
  • The stable association of virion with the triple-gene-block protein 3-based complex of Bamboo mosaic virus.
The stable association of virion with the triple-gene-block protein 3-based complex of Bamboo mosaic virus.
PLoS pathogens (2013-06-12)
Yuan-Lin Chou, Yi-Jing Hung, Yang-Hao Tseng, Hsiu-Ting Hsu, Jun-Yi Yang, Chiung-Hua Wung, Na-Sheng Lin, Menghsiao Meng, Yau-Heiu Hsu, Ban-Yang Chang
ABSTRACT
The triple-gene-block protein 3 (TGBp3) of Bamboo mosaic virus (BaMV) is an integral endoplasmic reticulum (ER) membrane protein which is assumed to form a membrane complex to deliver the virus intracellularly. However, the virus entity that is delivered to plasmodesmata (PD) and its association with TGBp3-based complexes are not known. Results from chemical extraction and partial proteolysis of TGBp3 in membrane vesicles revealed that TGBp3 has a right-side-out membrane topology; i.e., TGBp3 has its C-terminal tail exposed to the outer surface of ER. Analyses of the TGBp3-specific immunoprecipitate of Sarkosyl-extracted TGBp3-based complex revealed that TGBp1, TGBp2, TGBp3, capsid protein (CP), replicase and viral RNA are potential constituents of virus movement complex. Substantial co-fractionation of TGBp2, TGBp3 and CP, but not TGBp1, in the early eluted gel filtration fractions in which virions were detected after TGBp3-specific immunoprecipitation suggested that the TGBp2- and TGBp3-based complex is able to stably associate with the virion. This notion was confirmed by immunogold-labeling transmission electron microscopy (TEM) of the purified virions. In addition, mutational and confocal microscopy analyses revealed that TGBp3 plays a key role in virus cell-to-cell movement by enhancing the TGBp2- and TGBp3-dependent PD localization of TGBp1. Taken together, our results suggested that the cell-to-cell movement of potexvirus requires stable association of the virion cargo with the TGBp2- and TGBp3-based membrane complex and recruitment of TGBp1 to the PD by this complex.
Sigma-Aldrich
trans-2-Bromovinylboronic acid MIDA ester, technical grade
Pricing and availability is not currently available.