MilliporeSigma
FRET imaging.
Nature biotechnology (2003-11-05)
Elizabeth A Jares-Erijman, Thomas M Jovin
ABSTRACT
Förster (or Fluorescence) Resonance Energy Transfer (FRET) is unique in generating fluorescence signals sensitive to molecular conformation, association, and separation in the 1-10 nm range. We introduce a revised photophysical framework for the phenomenon and provide a systematic catalog of FRET techniques adapted to imaging systems, including new approaches proposed as suitable prospects for implementation. Applications extending from a single molecule to live cells will benefit from multidimensional microscopy techniques, particularly those adapted for optical sectioning and incorporating new algorithms for resolving the component contributions to images of complex molecular systems.
Product Description
Supelco
Atto 565 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Pricing and availability is not currently available.