MilliporeSigma
  • Localized-statistical quantification of human serum proteome associated with type 2 diabetes.
Localized-statistical quantification of human serum proteome associated with type 2 diabetes.
PloS one (2008-09-17)
Rong-Xia Li, Hai-Bing Chen, Kang Tu, Shi-Lin Zhao, Hu Zhou, Su-Jun Li, Jie Dai, Qing-Run Li, Song Nie, Yi-Xue Li, Wei-Ping Jia, Rong Zeng, Jia-Rui Wu
ABSTRACT
Recent advances in proteomics have shed light to discover serum proteins or peptides as biomarkers for tracking the progression of diabetes as well as understanding molecular mechanisms of the disease. In this work, human serum of non-diabetic and diabetic cohorts was analyzed by proteomic approach. To analyze total 1377 high-confident serum-proteins, we developed a computing strategy called localized statistics of protein abundance distribution (LSPAD) to calculate a significant bias of a particular protein-abundance between these two cohorts. As a result, 68 proteins were found significantly over-represented in the diabetic serum (p<0.01). In addition, a pathway-associated analysis was developed to obtain the overall pathway bias associated with type 2 diabetes, from which the significant over-representation of complement system associated with type 2 diabetes was uncovered. Moreover, an up-stream activator of complement pathway, ficolin-3, was observed over-represented in the serum of type 2 diabetic patients, which was further validated with statistic significance (p = 0.012) with more clinical samples. The developed LSPAD approach is well fit for analyzing proteomic data derived from biological complex systems such as plasma proteome. With LSPAD, we disclosed the comprehensive distribution of the proteins associated with diabetes in different abundance levels and the involvement of ficolin-related complement activation in diabetes.
Sigma-Aldrich
MISSION® esiRNA, targeting human FCN3
Pricing and availability is not currently available.