MilliporeSigma
  • A new method for the stereoselective synthesis of alpha- and beta-glycosylamines using the Burgess reagent.
A new method for the stereoselective synthesis of alpha- and beta-glycosylamines using the Burgess reagent.
Journal of the American Chemical Society (2004-05-20)
K C Nicolaou, Scott A Snyder, Annie Z Nalbandian, Deborah A Longbottom
ABSTRACT
Although glycosylamines constitute an important group of carbohydrates from the standpoint of biology and medicine, methods for their synthesis typically lack substrate generality and/or result in variable stereoselectivity, especially in complex contexts. In this communication, we report an operationally simple method for the synthesis of both alpha- and beta-glycosylamines using the Burgess reagent that overcomes many of these limitations in a bare minimum of synthetic steps.
Sigma-Aldrich
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide, inner salt, 97%
Pricing and availability is not currently available.