MilliporeSigma
Synthesis of fused heteroarylprolines and pyrrolopyrroles.
The Journal of organic chemistry (2004-07-03)
Guillaume Jeannotte, William D Lubell
ABSTRACT
Fused heteroarylprolines were prepared starting from 4-oxo-N-(PhF)proline benzyl ester (6, PhF = 9-(9-phenylfluorenyl)) following two approaches. First, allylation of oxoproline 6 followed by Wacker oxidation gave 1,4-dione 8 that was selectively converted to pyrroloproline 10b, pyrrolopyrrole 12, and pyridazinoproline 9. Second, aldol condensation of oxoproline 6 with a series of N-(Boc)-alpha-amino aldehydes 15a-e and acid-catalyzed cyclization gave pyrroloprolines 17a-e possessing a variety of pyrrole 5-position substituents. Conditions for the selective deprotection and alkylation of the pyrrole nitrogen of pyrroloprolines 17 were developed to expand the diversity of the heteoaryl systems. Finally, hydrogenolytic cleavage of the PhF and benzyl groups followed by subsequent protection with Boc, Fmoc, and Moz groups was performed to obtain analogues suitable for peptide synthesis. The enantiomeric purity of N-(Boc)pyrrolo-proline 21a was ascertained by coupling to l- and d-phenylalanine methyl ester and examination of the diastereotopic pyrrole protons, which demonstrated the dipeptides to be of >99% diastereomeric purity. These approaches have thus delivered the first series of enantiopure fused arylprolines for application as arylglycine-proline chimeras in structure-activity studies of biologically active compounds.
Sigma-Aldrich
N-Boc-2-aminoacetaldehyde, 95%
Sigma-Aldrich
tert-Butyl bromoacetate, 98%