MilliporeSigma
  • [Bis(2-methoxyethyl)amino]sulfur trifluoride, the Deoxo-Fluor reagent: application toward one-flask transformations of carboxylic acids to amides.
[Bis(2-methoxyethyl)amino]sulfur trifluoride, the Deoxo-Fluor reagent: application toward one-flask transformations of carboxylic acids to amides.
The Journal of organic chemistry (2004-03-31)
Jonathan M White, Ashok Rao Tunoori, Brandon J Turunen, Gunda I Georg
ABSTRACT
The use of the Deoxo-Fluor reagent is a versatile method for acyl fluoride generation and subsequent one-flask amide coupling. It provides mild conditions and facile purification of the desired products in good to excellent yields. We have explored the utility of this reagent for the one-flask conversion of acids to amides and Weinreb amides and as a peptide-coupling reagent.