MilliporeSigma
  • Total synthesis of TMC-95A and -B via a new reaction leading to Z-enamides. Some preliminary findings as to SAR.
Total synthesis of TMC-95A and -B via a new reaction leading to Z-enamides. Some preliminary findings as to SAR.
Journal of the American Chemical Society (2004-05-20)
Songnian Lin, Zhi-Qiang Yang, Benjamin H B Kwok, Michael Koldobskiy, Craig M Crews, Samuel J Danishefsky
ABSTRACT
A full account of the total syntheses of proteasome inhibitors TMC-95A and -B is provided. A key feature of the syntheses involved installation of a cis-propenylamide moiety by a thermal rearrangement of an alpha-silylallyl amide. The scope and mechanism of the enamide-forming reaction are discussed. Also provided are some preliminary results from SAR studies. It was found that simplified analogues can retain the full potency of proteasome inhibition.
Sigma-Aldrich
[1,1′-Bis(diphenylphosphino)ferrocene]dichloropalladium(II), complex with dichloromethane