MilliporeSigma
  • Mesoporous NiO crystals with dominantly exposed {110} reactive facets for ultrafast lithium storage.
Mesoporous NiO crystals with dominantly exposed {110} reactive facets for ultrafast lithium storage.
Scientific reports (2012-12-12)
Dawei Su, Mike Ford, Guoxiu Wang
ABSTRACT
Faceted crystals with exposed highly reactive planes have attracted intensive investigations for applications such as hydrogen production, enhanced catalytic activity, and electrochemical energy storage and conversion. Herein, we report the synthesis of mesoporous NiO crystals with dominantly exposed {110} reactive facets by the thermal conversion of hexagonal Ni(OH)(2) nanoplatelets. When applied as anode materials in lithium-ion batteries, mesoporous NiO crystals exhibit a high reversible lithium storage capacity of 700 mAh g(-1) at 1 C rate in 100 cycles and an excellent cyclability. In particular, the dominantly exposed {110} reactive facets and mesoporous nanostructure of NiO crystals lead to ultrafast lithium storage, which mimics the high power delivery of supercapacitors.