MilliporeSigma
  • 5-androstenediol ameliorates pleurisy, septic shock, and experimental autoimmune encephalomyelitis in mice.
5-androstenediol ameliorates pleurisy, septic shock, and experimental autoimmune encephalomyelitis in mice.
Autoimmune diseases (2010-12-29)
Ferdinando Nicoletti, Dominick L Auci, Katia Mangano, Jaime Flores-Riveros, Sonia Villegas, James M Frincke, Christopher L Reading, Halina Offner
ABSTRACT
Androstenediol (androst-5-ene-3β,17β-diol; 5-AED), a natural adrenal steroid, has been shown to suppress experimental autoimmune encephalomyelitis (EAE) in female SJL/J mice. We here report that 5-AED limits inflammation and proinflammatory cytokines including TNFα in murine models of carrageenan-induced pleurisy and lippopolysaccaride- (LPS) induced septic shock. 5-AED binds to and transactivates sex steroid receptors with the same general rank order of potency (ERβ > ERα ≫ AR). 5-AED provides benefit in EAE in a dose-dependent fashion, even when treatment is delayed until onset of disease. The minimally effective dose may be as low as 4 mg/kg in mice. However, benefit was not observed when 5-AED was given in soluble formulation, leading to a short half-life and rapid clearance. These observations suggest that treatment with 5-AED limits the production of pro-inflammatory cytokines in these animal models and, ultimately, when formulated and administered properly, may be beneficial for patients with multiple sclerosis and other Th1-driven autoimmune diseases.
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype enteritidis, purified by phenol extraction
Pricing and availability is not currently available.