- A fatty acyl-CoA oxidizing system in rat liver peroxisomes; enhancement by clofibrate, a hypolipidemic drug.
A fatty acyl-CoA oxidizing system in rat liver peroxisomes; enhancement by clofibrate, a hypolipidemic drug.
Proceedings of the National Academy of Sciences of the United States of America (1976-06-01)
P B Lazarow, C De Duve
PMID180535
ABSTRACT
Purified rat liver peroxisomes contain a cyanide-insensitive fatty acyl-CoA oxidizing system that uses O2 and NAD as electron acceptors. The system was detected by the ability of added palmitoyl-CoA to elicit O2 consumption, H2O2 production, and O2-dependent NAD reduction. The activity of this system is increased approximately one order of magnitude in rats treated with clofibrate, a hypolipidemic drug known to cause peroxisomal proliferation.