MilliporeSigma
  • N(6)-alkyl-2-alkynyl derivatives of adenosine as potent and selective agonists at the human adenosine A(3) receptor and a starting point for searching A(2B) ligands.
N(6)-alkyl-2-alkynyl derivatives of adenosine as potent and selective agonists at the human adenosine A(3) receptor and a starting point for searching A(2B) ligands.
Journal of medicinal chemistry (2002-07-12)
Rosaria Volpini, Stefano Costanzi, Catia Lambertucci, Sara Taffi, Sauro Vittori, Karl-Norbert Klotz, Gloria Cristalli
ABSTRACT
A series of N(6)-alkyl-2-alkynyl derivatives of adenosine (Ado) have been synthesized and evaluated for their affinity at human A(1), A(2A), and A(3) receptors and for their potency at A(2B) adenosine receptor subtypes. The corresponding 2-(1-alkynyl) derivatives of 5'-N-ethylcarboxamidoadenosine (NECA) and Ado are used as reference compounds. Binding studies demonstrated that the activities of 2-alkynylAdos were slightly increased for the adenosine A(1) receptor and slightly decreased for both A(3) and A(2B) subtypes compared to those of their corresponding NECA derivatives, whereas the A(2A) receptor affinities of the two series of nucleosides were similar. The presence of a methyl group on N(6) of the 2-alkynyladenosines, inducing an increase in affinity at the human A(3) receptor and a decrease at the other subtypes, resulted in an increase in A(3) selectivity. In particular, 2-phenylethynyl-N(6)-methylAdo (8b) showed an A(3) affinity in the low nanomolar range (K(i)(A(3)) = 3.4 nM), with a A(1)/A(3) and A(2A)/A(3) selectivity of about 500 and 2500, respectively. These findings motivated us to search for the preparation of new selective radioligands for the A(3) subtype; hence, a procedure to introduce a tritiated alkylamino group in these molecules was carried out. As far as the potency at the A(2B) receptor, the type of 2-alkynyl chain and the presence of the ethylcarboxamido group on the sugar seem to be very important; in fact, the (S)-2-phenylhydroxypropynylNECA [(S)-PHPNECA, 1e, EC(50)(A(2B)) = 0.22 microM] proved to be one of the most potent A(2B) agonist reported so far. On the other hand, the (S)-2-phenylhydroxypropynyl-N(6)-ethylAdo (9e, EC(50)(A(2B)) = 0.73 microM) showed a significantly increase of potency at the A(2B) subtype in comparison with the N(6)-methyl, N(6)-isopropyl, and the unsubstituted adenosine derivatives, although it resulted in being less potent than (S)-PHPNECA (1e, EC(50)(A(2B)) = 0.22 microM). These observations suggest that the introduction of an ethyl group in the N(6)-position and an ethylcarboxamido substituent in the 4'-position of (S)-2-phenylhydroxypropynyladenosine could lead to a compound endowed with high potency at the A(2B) receptor.