MilliporeSigma
  • Trough:peak ratio of the blood pressure response to angiotensin converting enzyme inhibitors.
Trough:peak ratio of the blood pressure response to angiotensin converting enzyme inhibitors.
Journal of hypertension. Supplement : official journal of the International Society of Hypertension (1994-11-01)
A Salvetti, L Di Venanzio, P Arrighi, F Arzilli
ABSTRACT
SHORT- VERSUS LONG-ACTING ANGIOTENSIN CONVERTING ENZYME (ACE) INHIBITORS: Although ACE inhibitors are widely used in the treatment of hypertension, there are few data on trough:peak ratios and the data are contradictory. Part of the explanation for this lies in differences in pharmacological properties. Depending on the kinetics of elimination, the trough:peak ratio of short- and long-acting ACE inhibitors has to be evaluated according to a dose regimen of twice or once a day, respectively, and must take account of the dose used, since long-acting ACE inhibitors appear to have a dose-dependent trough:peak effect. Further explanations for the contradictory trough:peak ratios reported for ACE inhibitors include measurement methods (clinic blood pressure versus ambulatory monitoring) and study design. TROUGH:PEAK RATIO: Data from randomly allocated, placebo-controlled studies indicate that both the short-acting ACE inhibitors captopril and quinapril given twice a day and the long-acting ACE inhibitors enalapril, lisinopril, benazepril and cilazapril given once a day have an acceptable trough:peak ratio (> 50%). The evidence suggests that when chemically different ACE inhibitors with similar kinetics of elimination are administered at equipotent doses, similar trough:peak ratios are obtained.
Sigma-Aldrich
Benazepril hydrochloride, ≥98% (HPLC), solid
Pricing and availability is not currently available.
Sigma-Aldrich
Quinapril hydrochloride, ≥98% (HPLC), solid
Pricing and availability is not currently available.