MilliporeSigma
  • Quantitative determination of chitosan and the percentage of free amino groups.
Quantitative determination of chitosan and the percentage of free amino groups.
Analytical biochemistry (1993-06-01)
E Curotto, F Aros
ABSTRACT
Chitosans were quantified with ninhydrin, a reagent normally used for recognizing and quantifying amino groups. The reaction was time dependent and there was no effect when different acids were added to the mixture. This method was used to determine the percentage of free amino groups in chitosans of different origins.
Sigma-Aldrich
Ninhydrin, ACS reagent
Pricing and availability is not currently available.