MilliporeSigma
  • Synthesis and cardiotonic activity of 2-substituted 5-cyano-1,6-dihydro-6-oxo-3-pyridinecarboxylic acids and their methyl or ethyl esters.
Synthesis and cardiotonic activity of 2-substituted 5-cyano-1,6-dihydro-6-oxo-3-pyridinecarboxylic acids and their methyl or ethyl esters.
Farmaco (Societa chimica italiana : 1989) (1992-04-01)
L Mosti, G Menozzi, P Schenone, P Dorigo, R M Gaion, P Belluco
ABSTRACT
The synthesis of ethyl or methyl esters of 5-cyano-1,6-dihydro-6-oxo-3- pyridinecarboxylic acids carrying as 2-substituent a polar group such as CO2C2H5, (CH2)2CO2CH3, (CH2)3CO2C2H5, CH2OCH3, or CF3 group is described. Also 2-[5-cyano-1,6-dihydro-2-(1,1-dimethylethyl)-6-oxo-3-pyridyl]-2- oxoacetic acid and 2,5,6,8-tetrahydro-2,5-dioxo-1H-thiopyrano[3,4-b]pyridine-3-carbon itrile were prepared. Nearly all the above esters gave routinely the corresponding carboxylic acids by alkaline hydrolysis followed by acidification. As milrinone analogues, the above compounds were tested on contractile activity and frequency rate of spontaneously beating atria from reserpine-treated guinea-pigs. 5-Cyano-2-trifluoromethyl-1,6- dihydro-6-oxo-3-pyridinecarboxylic acid and, in a lesser degree, the relative ethyl ester showed an appreciable positive inotropic activity, although inferior to that of milrinone.
Sigma-Aldrich
Ethyl 2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate,mixture of cis and trans, 96%