MilliporeSigma
  • Conformational studies of human plasminogen and plasminogen fragments: evidence for a novel third conformation of plasminogen.
Conformational studies of human plasminogen and plasminogen fragments: evidence for a novel third conformation of plasminogen.
Biochemistry (1994-03-29)
J M Marshall, A J Brown, C P Ponting
ABSTRACT
The conformations of Glu-plasminogen and defined proteolytic fragments, in the presence and absence of 6-aminohexanoic acid (6-AHA), trans-4-(aminomethyl)cyclohexanecarboxylic acid (t-AMCHA), and benzamidine, were studied using three methods: size-exclusion high-performance liquid chromatography (SE-HPLC), small-angle X-ray scattering (SAXS), and dynamic laser light scattering (DLLS). The well-documented conformational change of Glu-plasminogen with 6-AHA or t-AMCHA was measured as a decrease in molecular elution time by SE-HPLC (8.93 +/- 0.01 to 8.32 +/- 0.01 min) and increases in radius of gyration (30.7 +/- 0.1 to 49.8 +/- 0.3 A) and Stokes radius (40.6 +/- 0.3 to 48.5 +/- 0.3 A) by SAXS and DLLS, respectively. The addition of benzamidine to Glu-plasminogen resulted in a conformation (radius of gyration 41.0 +/- 0.4 A and Stokes radius 46.6 +/- 0.3 A) distinct from that in the presence of 6-AHA. 6-AHA, but not benzamidine, induced significant conformational changes in Lys-plasminogen and kringles 1 + 2 + 3 + 4 + 5. We conclude that Glu-plasminogen adopts three distinct conformations involving two intramolecular interactions: one mediated by regions of the NH2-terminal peptide and kringle 5, competed for by 6-AHA or benzamidine, and the other possibly between kringles 3 and 4, competed for by 6-AHA but not benzamidine.
Sigma-Aldrich
trans-4-(Aminomethyl)cyclohexanecarboxylic acid, 97%
Pricing and availability is not currently available.