MilliporeSigma
  • Regulation of amino acid transporter trafficking by mTORC1 in primary human trophoblast cells is mediated by the ubiquitin ligase Nedd4-2.
Regulation of amino acid transporter trafficking by mTORC1 in primary human trophoblast cells is mediated by the ubiquitin ligase Nedd4-2.
Clinical science (London, England : 1979) (2015-11-27)
Fredrick J Rosario, Kris Genelyn Dimasuay, Yoshikatsu Kanai, Theresa L Powell, Thomas Jansson
ABSTRACT
Changes in placental amino acid transfer directly contribute to altered fetal growth, which increases the risk for perinatal complications and predisposes for the development of obesity, diabetes and cardiovascular disease later in life. Placental amino acid transfer is critically dependent on the expression of specific transporters in the plasma membrane of the trophoblast, the transporting epithelium of the human placenta. However, the molecular mechanisms regulating this process are largely unknown. Nedd4-2 is an ubiquitin ligase that catalyses the ubiquitination of proteins, resulting in proteasomal degradation. We hypothesized that inhibition of mechanistic target of rapamycin complex 1 (mTORC1) decreases amino acid uptake in primary human trophoblast (PHT) cells by activation of Nedd4-2, which increases transporter ubiquitination resulting in decreased transporter expression in the plasma membrane. mTORC 1 inhibition increased the expression of Nedd4-2, promoted ubiquitination and decreased the plasma membrane expression of SNAT2 (an isoform of the System A amino acid transporter) and LAT1 (a System L amino acid transporter isoform), resulting in decreased cellular amino acid uptake. Nedd4-2 silencing markedly increased the trafficking of SNAT2 and LAT1 to the plasma membrane, which stimulated cellular amino acid uptake. mTORC1 inhibition by silencing of raptor failed to decrease amino acid transport following Nedd4-2 silencing. In conclusion, we have identified a novel link between mTORC1 signalling and ubiquitination, a common posttranslational modification. Because placental mTORC1 is inhibited in fetal growth restriction and activated in fetal overgrowth, we propose that regulation of placental amino acid transporter ubiquitination by mTORC1 and Nedd4-2 constitutes a molecular mechanisms underlying abnormal fetal growth.