MilliporeSigma
  • Contributing effect of various washing procedures and additives on the decline pattern of diethofencarb in crown daisy, a model of leafy vegetables.
Contributing effect of various washing procedures and additives on the decline pattern of diethofencarb in crown daisy, a model of leafy vegetables.
Food chemistry (2016-02-13)
Sung-Woo Kim, A M Abd El-Aty, Jeong-Heui Choi, Young-Jun Lee, Truong T B Lieu, Hyung Suk Chung, Md Musfiqur Rahman, Ok-Ja Choi, Ho-Chul Shin, Gyu-Seek Rhee, Moon-Ik Chang, Hee Jung Kim, Jae-Han Shim
ABSTRACT
The effects of various washing procedures, including stagnant, running, and stagnant and running tap water, and the use of washing solutions and additives, namely NaCl (1% and 2%), vinegar (2%, 5%, and 10%), detergent (0.5% and 1%), and charcoal (1% and 2%), on the reduction rate of diethofencarb were estimated in field-incurred crown daisy, a model of leafy vegetables, grown under greenhouses located in 3 different areas (Gwangju, Naju, and Muan). The original Quick, Easy, Cheap, Effective, Rugged, and Safe "QuEChERS" method was modified for extraction and liquid chromatography-tandem mass spectrometry (LC/MS/MS) was used for analysis. The recovery of diethofencarb in unwashed and washed samples was satisfactory and ranged between 84.28% and 115.32% with relative standard deviations (RSDs) of <6%. The residual levels decreased following washing with stagnant, running, and stagnant+running tap water (i.e., decline in levels increased from 65.08% to 85.02%, 69.99 to 86.79, and 74.75 to 88.96, respectively). The percentage of decline increased and ranged from 77.46% to 91.19% following washing with various solutions. Application of 1% detergent was found to be the most effective washing method for reducing the residues in crown daisy. Additionally, washing with stagnant and running tap water or even stagnant water for 5 min might reduce the residue levels substantially, making the prepared food safe for human consumption.
Product Description
Supelco
Diethofencarb, PESTANAL®, analytical standard