MilliporeSigma
  • Inverse electron demand Diels-Alder reactions of 1,2,3-triazines: pronounced substituent effects on reactivity and cycloaddition scope.
Inverse electron demand Diels-Alder reactions of 1,2,3-triazines: pronounced substituent effects on reactivity and cycloaddition scope.
Journal of the American Chemical Society (2011-07-09)
Erin D Anderson, Dale L Boger
ABSTRACT
A systematic study of the inverse electron demand Diels-Alder reactions of 1,2,3-triazines is disclosed, including an examination of the impact of a C5 substituent. Such substituents were found to exhibit a remarkable impact on the cycloaddition reactivity of the 1,2,3-triazine without altering, and perhaps even enhancing, the intrinsic cycloaddition regioselectivity. The study revealed not only that the reactivity may be predictably modulated by a C5 substituent (R = CO(2)Me > Ph > H) but also that the impact is of a magnitude to convert 1,2,3-triazine (1) and its modest cycloaddition scope into a heterocyclic azadiene system with a reaction scope that portends extensive synthetic utility, expanding the range of participating dienophiles. Significantly, the studies define a now powerful additional heterocyclic azadiene, complementary to the isomeric 1,2,4-triazines and 1,3,5-triazines, capable of dependable participation in inverse electron demand Diels-Alder reactions, extending the number of complementary heterocyclic ring systems accessible with implementation of the methodology.
Sigma-Aldrich
5-phenyl-1,2,3-triazine, ≥95%