MilliporeSigma
  • Chemical modification of silk sericin in lithium chloride/dimethyl sulfoxide solvent with 4-cyanophenyl isocyanate.
Chemical modification of silk sericin in lithium chloride/dimethyl sulfoxide solvent with 4-cyanophenyl isocyanate.
Biomacromolecules (2004-07-13)
Hidetoshi Teramoto, Ken-ichi Nakajima, Chiyuki Takabayashi
ABSTRACT
This paper reports chemical modification of silk sericin in LiCl/dimethyl sulfoxide (DMSO) solvent with 4-cyanophenyl isocyanate. Sericin is a highly hydrophilic protein secreted by Bombyx mori, serving as a protein glue in a cocoon. LiCl/DMSO was found to be a good solvent of sericin and useful for homogeneous modification of its abundant hydroxyl groups under nonaqueous condition. Fourier transform infrared (FTIR) analysis of the modified sericins revealed that 4-cyanophenyl groups were incorporated into sericin molecules mainly through urethane linkages. Several characteristics of the modified sericins such as solubility characteristic, hygroscopic property, and thermal stability were investigated. Secondary structure analysis using FTIR spectra suggested that formation of strong intermolecular hydrogen bonds was inhibited by the modification that is probably attributable to the incorporation of bulky 4-cyanophenyl groups. These results demonstrate that chemical modification of sericin using LiCl/DMSO solvent markedly alters its characteristics.
Sigma-Aldrich
4-Cyanophenyl isocyanate, 97%
Pricing and availability is not currently available.