Merck
  • MicroRNA-31 initiates lung tumorigenesis and promotes mutant KRAS-driven lung cancer.
MicroRNA-31 initiates lung tumorigenesis and promotes mutant KRAS-driven lung cancer.
The Journal of clinical investigation (2015-12-15)
Mick D Edmonds, Kelli L Boyd, Tamara Moyo, Ramkrishna Mitra, Robert Duszynski, Maria Pia Arrate, Xi Chen, Zhongming Zhao, Timothy S Blackwell, Thomas Andl, Christine M Eischen
ABSTRACT
MicroRNA (miR) are important regulators of gene expression, and aberrant miR expression has been linked to oncogenesis; however, little is understood about their contribution to lung tumorigenesis. Here, we determined that miR-31 is overexpressed in human lung adenocarcinoma and this overexpression independently correlates with decreased patient survival. We developed a transgenic mouse model that allows for lung-specific expression of miR-31 to test the oncogenic potential of miR-31 in the lung. Using this model, we observed that miR-31 induction results in lung hyperplasia, followed by adenoma formation and later adenocarcinoma development. Moreover, induced expression of miR-31 in mice cooperated with mutant KRAS to accelerate lung tumorigenesis. We determined that miR-31 regulates lung epithelial cell growth and identified 6 negative regulators of RAS/MAPK signaling as direct targets of miR-31. Our study distinguishes miR-31 as a driver of lung tumorigenesis that promotes mutant KRAS-mediated oncogenesis and reveals that miR-31 directly targets and reduces expression of negative regulators of RAS/MAPK signaling.
Sigma-Aldrich
Anti-Spred-2 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Pricing and availability is not currently available.