MilliporeSigma
  • Independent Control of Topography for 3D Patterning of the ECM Microenvironment.
Independent Control of Topography for 3D Patterning of the ECM Microenvironment.
Advanced materials (Deerfield Beach, Fla.) (2015-11-10)
Jiyun Kim, Jack Rory Staunton, Kandice Tanner
ABSTRACT
Biomimetic extracellular matrix (ECM) topographies driven by the magnetic-field-directed self-assembly of ECM protein-coated magnetic beads are fabricated. This novel bottom-up method allows us to program isotropic, anisotropic, and diverse hybrid ECM patterns without changing other physicochemical properties of the scaffold material. It is demonstrated that this 3D anisotropic matrix is able to guide the dendritic protrusion of cells.
Sigma-Aldrich
Nerve Growth Factor-7S from murine submaxillary gland, NGF-7S, lyophilized powder, suitable for cell culture
Sigma-Aldrich
N-Acetyl-D-lactosamine, ≥98% (TLC)
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 150,000