MilliporeSigma
Heat shock protein 70-1A is a novel angiogenic regulator.
Biochemical and biophysical research communications (2015-12-15)
Taek-Keun Kim, Hee Jun Na, Woo Ran Lee, Mee Hyun Jeoung, Sukmook Lee
ABSTRACT
Heat shock protein 70-1A (HSP70-1A) is a stress-inducible protein that provides an essential intracellular molecular chaperone function; however, the mechanism of HSP70-1A in angiogenesis has not been clarified. Herein, HSP70-1A gene silencing implicated this protein in angiogenesis. Additionally, recombinant human HSP70-1A (rhHSP70-1A) was able to stimulate human umbilical vein endothelial cell (HUVEC) migration and tube formation in vitro and microvessel formation in vivo similarly to recombinant human vascular endothelial growth factor (rhVEGF). Furthermore, rhHSP70-1A was tightly bound to the surface of HUVECs and participated in extracellular signal-related kinase (ERK)-dependent angiogenesis. Together, these results implicate HSP70-1A as a novel angiogenic regulator.
Sigma-Aldrich
MISSION® esiRNA, targeting human HSPA1A (2)