MilliporeSigma
  • [Association of -63A/C polymorphism of glutathione S-transferase M3 gene with essential hypertension in Chinese population].
[Association of -63A/C polymorphism of glutathione S-transferase M3 gene with essential hypertension in Chinese population].
Zhonghua yi xue yi chuan xue za zhi = Zhonghua yixue yichuanxue zazhi = Chinese journal of medical genetics (2007-10-09)
Jian Liu, Miao Li, Man-long Qi, Xiao-liang Liu, Ying-hui Li, Ling-yu Fu, Yan-yan Zhao
ABSTRACT
To investigate the distribution of -63A/C polymorphism of human glutathione S-transferase M3(GSTM3) gene in Chinese Han population and the association of -63A/C polymorphism with essential hypertension (EH). -63A/C polymorphism of GSTM3 gene was examined by polymerase chain reaction-restriction fragment length polymorphism (PCR-RFLP) in 234 patients with EH and 328 healthy controls. The genotypes were confirmed partially by DNA sequencing. The distribution of GSTM3 -63A/C was in Hardy-Weinberg equilibrium. The CC genotype frequency in EH group (6%) was significantly higher than that in control group(1.8%) (P < 0.05). After adjustment for age, body mass index, and other risk factors, the CC genotype was independently associated with hypertension (OR=3.447, 95%CI: 1.19-7.63; P=0.04). The GSTM3 -63A/C polymorphism was associated with EH in Chinese Han population, the C allele might be a risk factor for EH in Chinese Han nationality.