MilliporeSigma
  • Measurement of succinyl-carnitine and methylmalonyl-carnitine on dried blood spot by liquid chromatography-tandem mass spectrometry.
Measurement of succinyl-carnitine and methylmalonyl-carnitine on dried blood spot by liquid chromatography-tandem mass spectrometry.
Clinica chimica acta; international journal of clinical chemistry (2013-11-26)
Cristiano Rizzo, Sara Boenzi, Rita Inglese, Giancarlo la Marca, Maurizio Muraca, Tegra Barreiro Martinez, David W Johnson, Eleonora Zelli, Carlo Dionisi-Vici
ABSTRACT
Methylmalonic aciduria (MMA) is one of the most frequent organic acidurias, a class of diseases caused by enzymatic defects mainly involved in the catabolism of branched-chain amino acids. Recently, mild MMA and C4-dicarboxylyl-carnitine (C4DC-C) accumulation have been reported in patients carrying mutation in genes encoding the α-subunit (SUCLG1) and the β-subunit (SUCLA2) of the ADP-forming succinyl-CoA synthetase (SCS). We developed a liquid chromatography-tandem mass spectrometry (LC-MS/MS) method to quantify in dried blood spot the two isobaric compounds of C4DC-C, succinyl-carnitine and methylmalonyl-carnitine, to allow the differential diagnosis between classical MMA and SCS-related defects. This method, with an easy liquid-phase extraction and derivatization procedure, has been validated to demonstrate the specificity, linearity, recovery, lowest limit of quantification (LLOQ), accuracy and precision for quantitative determination of blood succinyl-carnitine and methylmalonyl-carnitine. The assay was linear over a concentration range of 0.025-10 μmol/L and achieved the LLOQ of 0.025 μmol/L for both metabolites. The average slope, intercept, and coefficient of linear regression (r(2)) were respectively: 0.3389 (95% confidence interval 0.2888-0.3889), 0.0113 (95% confidence interval -0.0157 to 0.0384), 0.9995 (95% confidence interval 0.9990-1.0000) for succinyl-carnitine and 0.5699 (95% confidence interval 0.5263-0.6134), 0.0319 (95% confidence interval -0.0038 to 0.0677), 0.9997 (95% confidence interval 0.9995-1.0000) for methylmalonyl-carnitine. Within-day and between-day coefficients of variation (CV) were 1.94% and 3.19% for succinyl-carnitine and 3.21%, and 2.56 for methylmalonyl-carnitine. This method is accurate and provides a new tool to differentiate patients with classical methylmalonic acidemia from those with SCS-related defects.
Product Description
Supelco
O-Succinyl-L-carnitine lithium salt, analytical standard