MilliporeSigma
  • PcpA, which is involved in the degradation of pentachlorophenol in Sphingomonas chlorophenolica ATCC39723, is a novel type of ring-cleavage dioxygenase.
PcpA, which is involved in the degradation of pentachlorophenol in Sphingomonas chlorophenolica ATCC39723, is a novel type of ring-cleavage dioxygenase.
FEBS letters (1999-10-20)
Y Ohtsubo, K Miyauchi, K Kanda, T Hatta, H Kiyohara, T Senda, Y Nagata, Y Mitsui, M Takagi
ABSTRACT
The pentachlorophenol (PCP) mineralizing bacterium Sphingomonas chlorophenolica ATCC39723 degrades PCP via 2,6-dichlorohydroquinone (2,6-DCHQ). The pathway converting PCP to 2,6-DCHQ has been established previously; however, the pathway beyond 2,6-DCHQ is not clear, although it has been suggested that a PcpA plays a role in 2, 6-DCHQ conversion. In this study, PcpA expressed in Escherichia coli was purified to homogeneity and shown to have novel ring-cleavage dioxygenase activity in conjunction with hydroquinone derivatives, and converting 2,6-DCHQ to 2-chloromaleylacetate.