MilliporeSigma
  • Non-Instrumented Nucleic Acid Amplification (NINA) for Rapid Detection of
Non-Instrumented Nucleic Acid Amplification (NINA) for Rapid Detection of
Biological engineering transactions (2011-01-01)
Ryo Kubota, Paul LaBarre, Jered Singleton, Andy Beddoe, Bernhard H Weigl, Anne M Alvarez, Daniel M Jenkins
ABSTRACT
We report on the use of a non-instrumented device for the implementation of a loop-mediated amplification (LAMP) based assay for the select-agent bacterial-wilt pathogen