MilliporeSigma
  • Activation of IP3 receptors requires an endogenous 1-8-14 calmodulin-binding motif.
Activation of IP3 receptors requires an endogenous 1-8-14 calmodulin-binding motif.
The Biochemical journal (2012-09-27)
Yi Sun, Ana M Rossi, Taufiq Rahman, Colin W Taylor
ABSTRACT
Binding of IP3 (inositol 1,4,5-trisphosphate) to the IP3-binding core (residues 224-604) of IP3Rs (IP3 receptors) initiates opening of these ubiquitous intracellular Ca2+ channels. The mechanisms are unresolved, but require conformational changes to pass through the suppressor domain (residues 1-223). A calmodulin-binding peptide derived from myosin light chain kinase uncouples these events. We identified a similar conserved 1-8-14 calmodulin-binding motif within the suppressor domain of IP3R1 and, using peptides and mutagenesis, we demonstrate that it is essential for IP3R activation, whether assessed by IP3-evoked Ca2+ release or patch-clamp recoding of nuclear IP3R. Mimetic peptides specifically inhibit activation of IP3R by uncoupling the IP3-binding core from the suppressor domain. Mutations of key hydrophobic residues within the endogenous 1-8-14 motif mimic the peptides. Our results show that an endogenous 1-8-14 motif mediates conformational changes that are essential for IP3R activation. The inhibitory effects of calmodulin and related proteins may result from disruption of this essential interaction.