MilliporeSigma
  • Ph(i-PrO)SiH2: An Exceptional Reductant for Metal-Catalyzed Hydrogen Atom Transfers.
Ph(i-PrO)SiH2: An Exceptional Reductant for Metal-Catalyzed Hydrogen Atom Transfers.
Journal of the American Chemical Society (2016-03-18)
Carla Obradors, Ruben M Martinez, Ryan A Shenvi
ABSTRACT
We report the discovery of an outstanding reductant for metal-catalyzed radical hydrofunctionalization reactions. Observations of unexpected silane solvolysis distributions in the HAT-initiated hydrogenation of alkenes reveal that phenylsilane is not the kinetically preferred reductant in many of these transformations. Instead, isopropoxy(phenyl)silane forms under the reaction conditions, suggesting that alcohols function as important silane ligands to promote the formation of metal hydrides. Study of its reactivity showed that isopropoxy(phenyl)silane is an exceptionally efficient stoichiometric reductant, and it is now possible to significantly decrease catalyst loadings, lower reaction temperatures, broaden functional group tolerance, and use diverse, aprotic solvents in iron- and manganese-catalyzed hydrofunctionalizations. As representative examples, we have improved the yields and rates of alkene reduction, hydration, hydroamination, and conjugate addition. Discovery of this broadly applicable, chemoselective, and solvent-versatile reagent should allow an easier interface with existing radical reactions. Finally, isotope-labeling experiments rule out the alternative hypothesis of hydrogen atom transfer from a redox-active β-diketonate ligand in the HAT step. Instead, initial HAT from a metal hydride to directly generate a carbon-centered radical appears to be the most reasonable hypothesis.
Sigma-Aldrich
Isopropoxy(phenyl)silane
Pricing and availability is not currently available.