MilliporeSigma
  • Efficacy of a combined inactivated porcine reproductive and respiratory syndrome virus vaccine using North American and European strains in specific pathogen free pigs.
Efficacy of a combined inactivated porcine reproductive and respiratory syndrome virus vaccine using North American and European strains in specific pathogen free pigs.
Veterinary journal (London, England : 1997) (2015-04-30)
Minjoo Yeom, Kwang-Soo Lyoo, Bo-Kyu Kang, Daesub Song, Bongkyun Park
ABSTRACT
In Korea, porcine reproductive and respiratory syndrome (PRRS) is caused by European (type 1) and North American (type 2) strains of PRRS virus (PRRSV). In the present study, the efficacy of a multi-strain PRRSV vaccine inactivated with binary ethylenimine (BEI) was evaluated in pigs. The vaccine contained one type 1 strain (GCEU0907) and two type 2 strains (GC4019 and GC6262). Three vaccinated groups (four pigs per group) and three mock vaccinated groups (four pigs per group) were challenged with infectious PRRSV (strains GC4019, GC6262 or GCEU0907), then euthanased at 28 days post-infection. Mean anti-PRRSV neutralising antibody titres were significantly higher in the vaccinated groups than in the mock vaccinated groups. Mean blood virus titres in the mock vaccinated groups were significantly higher than those in the vaccinated groups from 5 to 28 days post-infection. On pathological examination, there were less severe macroscopic and microscopic lesions in vaccinated pigs compared with mock vaccinated pigs.
Sigma-Aldrich
2-Bromoethylamine hydrobromide, 99%