MilliporeSigma
  • L-Quisqualic acid transport into hippocampal neurons by a cystine-sensitive carrier is required for the induction of quisqualate sensitization.
L-Quisqualic acid transport into hippocampal neurons by a cystine-sensitive carrier is required for the induction of quisqualate sensitization.
Neuroscience (2001-09-22)
L A Chase, R J Roon, L Wellman, A J Beitz, J F Koerner
ABSTRACT
A brief exposure of hippocampal slices to L-quisqualic acid sensitizes CA1 pyramidal neurons 30-250-fold to depolarization by two classes of excitatory amino acid analogues: (1) those whose depolarizing effects are rapidly terminated following washout, e.g. L-2-amino-4-phosphonobutanoic acid (L-AP4) and L-2-amino-6-phosphonohexanoic acid (L-AP6) and (2) those whose depolarizing effects persist following washout, e.g. L-aspartate-beta-hydroxamate (L-AbetaH). This process has been termed quisqualate sensitization. In this study we directly examine the role of amino acid transport systems in the induction of quisqualate sensitization. We report that L-quisqualate is a low-affinity substrate (K(M)=0.54 mM) for a high capacity (V(max)=0.9 nmol (mg protein)(-1) min(-1)) Na(+)-dependent transport system(s) and a high-affinity substrate (K(M)=0.033 mM) for a low-capacity (V(max)=0.051 nmol (mg protein)(-1) min(-1)) transporter with properties similar to the cystine/glutamate exchange carrier, System x(c-). We present evidence that suggests that System x(c-) participates in quisqualate sensitization. First, simultaneous application of L-quisqualate and inhibitors of System x(c-), but not inhibitors of Na(+)-dependent glutamate transporters, prevents the subsequent sensitization of hippocampal neurons to phosphonates or L-AbetaH. Second, L-quisqualic acid only sensitizes hippocampal neurons to other substrates of System x(c-), including cystine. Third, immunocytochemical analysis of L-quisqualate uptake demonstrates that only inhibitors of System x(c-) inhibit the highly concentrative uptake of L-quisqualate into a widely dispersed group of GABAergic hippocampal interneurons. We conclude that quisqualate sensitization is a direct consequence of the unique interaction of various excitatory amino acids, namely L-quisqualate, cystine, and phosphonates, with the exchange carrier, System x(c-). Therefore, the results of this study have important implications for the mechanism by which L-quisqualate, and other substrates of this transporter which are also excitatory amino acid agonists (such as glutamate and beta-N-oxalyl-L-alpha,beta-diaminopropionic acid, beta-L-ODAP) may trigger neurotoxicity.