MilliporeSigma
  • Comparison of neutrophil infiltration between type 1 and type 2 autoimmune pancreatitis.
Comparison of neutrophil infiltration between type 1 and type 2 autoimmune pancreatitis.
Pancreatology : official journal of the International Association of Pancreatology (IAP) ... [et al.] (2015-03-31)
Toshiyuki Mitsuyama, Kazushige Uchida, Kimi Sumimoto, Yuri Fukui, Tsukasa Ikeura, Toshiro Fukui, Akiyoshi Nishio, Nobuaki Shikata, Yoshiko Uemura, Sohei Satoi, Nobumasa Mizuno, Kenji Notohara, Tooru Shimosegawa, Giuseppe Zamboni, Luca Frulloni, Kazuichi Okazaki
ABSTRACT
Characteristics of type 2 autoimmune pancreatitis (AIP) is granulocyte epithelial lesions, called idiopathic duct-centric pancreatitis (IDCP). To clarify pathogenesis of IDCP, we investigated mechanism of neutrophil infiltration in type 1 AIP, called lymphoplasmacytic sclerosing pancreatitis (LPSP) and IDCP. This study was performed on resected pancreata from patients with alcoholic chronic pancreatitis (ACP, n = 10), LPSP (n = 10) and IDCP (n = 12). The number of neutrophils around the pancreatic ducts was counted. The expression of neutrophils chemoattractants granulocyte chemotactic protein-2 (GCP-2) and interleukin-8 (IL-8) in the pancreatic duct epithelia was examined using immunohistochemistry. The cell staining intensity is scored as negative (0), weak (1), moderate (2) or strong (3). The median number of neutrophils around the interlobular pancreatic ducts was significantly higher in IDCP (15.16; interquartile range [IQR]: 9.74-18.41) than in ACP (2.66; IQR: 1.33-4.33) (P < 0.05) and LPSP (3.16; IQR: 2.74-4.57) (P < 0.01). There was no significant difference in the median number of neutrophils around the intralobular pancreatic ducts among ACP (1.16; IQR: 0.33-3.41), LPSP (3.16; IQR: 0.74-5.5) and IDCP (3.00; IQR: 1.08-7.91). The median score of GCP-2 in the interlobular pancreatic duct epithelia was significantly higher in IDCP (1.5; IQR: 0.25-2) than in ACP (0; IQR: 0-0.75) (P < 0.05) and LPSP (0; IQR: 0-0.75) (P < 0.05). There was no significant difference in the median score of IL-8 in the interlobular pancreatic duct epithelia among ACP (0; IQR: 0-0.75), LPSP (1; IQR: 0-1.75) and IDCP (0.5; IQR: 0-1). Significantly increased neutrophil infiltration around the interlobular pancreatic duct in IDCP may depend on GCP-2.
Sigma-Aldrich
MISSION® esiRNA, targeting human CXCL6
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human CXCL5
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human TUBGCP2
Pricing and availability is not currently available.