MilliporeSigma
  • Different STAT Transcription Complexes Drive Early and Delayed Responses to Type I IFNs.
Different STAT Transcription Complexes Drive Early and Delayed Responses to Type I IFNs.
Journal of immunology (Baltimore, Md. : 1950) (2015-05-29)
Ali A Abdul-Sater, Andrea Majoros, Courtney R Plumlee, Stuart Perry, Ai Di Gu, Carolyn Lee, Sujan Shresta, Thomas Decker, Christian Schindler
ABSTRACT
IFNs, which transduce pivotal signals through Stat1 and Stat2, effectively suppress the replication of Legionella pneumophila in primary murine macrophages. Although the ability of IFN-γ to impede L. pneumophila growth is fully dependent on Stat1, IFN-αβ unexpectedly suppresses L. pneumophila growth in both Stat1- and Stat2-deficient macrophages. New studies demonstrating that the robust response to IFN-αβ is lost in Stat1-Stat2 double-knockout macrophages suggest that Stat1 and Stat2 are functionally redundant in their ability to direct an innate response toward L. pneumophila. Because the ability of IFN-αβ to signal through Stat1-dependent complexes (i.e., Stat1-Stat1 and Stat1-Stat2 dimers) has been well characterized, the current studies focus on how Stat2 is able to direct a potent response to IFN-αβ in the absence of Stat1. These studies reveal that IFN-αβ is able to drive the formation of a Stat2 and IFN regulatory factor 9 complex that drives the expression of a subset of IFN-stimulated genes, but with substantially delayed kinetics. These observations raise the possibility that this pathway evolved in response to microbes that have devised strategies to subvert Stat1-dependent responses.
Sigma-Aldrich
2-Hydroxypyridine, 97%