MilliporeSigma
  • Targeting interleukin-2 to the bone marrow stroma for therapy of acute myeloid leukemia relapsing after allogeneic hematopoietic stem cell transplantation.
Targeting interleukin-2 to the bone marrow stroma for therapy of acute myeloid leukemia relapsing after allogeneic hematopoietic stem cell transplantation.
Cancer immunology research (2015-02-13)
Christoph Schliemann, Katrin L Gutbrodt, Andrea Kerkhoff, Michele Pohlen, Stefanie Wiebe, Gerda Silling, Linus Angenendt, Torsten Kessler, Rolf M Mesters, Leonardo Giovannoni, Michael Schäfers, Bianca Altvater, Claudia Rossig, Inga Grünewald, Eva Wardelmann, Gabriele Köhler, Dario Neri, Matthias Stelljes, Wolfgang E Berdel
ABSTRACT
The antibody-based delivery of IL2 to extracellular targets expressed in the easily accessible tumor-associated vasculature has shown potent antileukemic activity in xenograft and immunocompetent murine models of acute myelogenous leukemia (AML), especially in combination with cytarabine. Here, we report our experience with 4 patients with relapsed AML after allogeneic hematopoietic stem cell transplantation (allo-HSCT), who were treated with the immunocytokine F16-IL2, in combination with low-dose cytarabine. One patient with disseminated extramedullary AML lesions achieved a complete metabolic response identified by PET/CT, which lasted 3 months. Two of 3 patients with bone marrow relapse achieved a blast reduction with transient molecular negativity. One of the 2 patients enjoyed a short complete remission before AML relapse occurred 2 months after the first infusion of F16-IL2. In line with a site-directed delivery of the cytokine, F16-IL2 led to an extensive infiltration of immune effector cells in the bone marrow. Grade 2 fevers were the only nonhematologic side effects in 2 patients. Grade 3 cytokine-release syndrome developed in the other 2 patients but was manageable in both cases with glucocorticoids. The concept of specifically targeting IL2 to the leukemia-associated stroma deserves further evaluation in clinical trials, especially in patients who relapse after allo-HSCT.
Product Description
Roche
Interleukin-2, human (hIL-2), recombinant (E. coli)