MilliporeSigma
  • Biochemical characterization of three distinct polygalacturonases from Neosartorya fischeri P1.
Biochemical characterization of three distinct polygalacturonases from Neosartorya fischeri P1.
Food chemistry (2015-06-05)
Xia Pan, Ke Li, Rui Ma, Pengjun Shi, Huoqing Huang, Peilong Yang, Kun Meng, Bin Yao
ABSTRACT
Polygalacturonase is one of the most important industrial pectinases. To enrich the genetic resources and develop new enzyme candidates, three polygalacturonase genes (Nfpg I-III) of glycosyl hydrolase family 28 were cloned from Neosartorya fischeri P1 and functionally expressed in Pichia pastoris. The purified recombinant proteins exhibited some distinguished properties. In comparison with other counterparts, NfPG I showed the highest specific activity (40, 123 U/mg), NfPG II had the highest temperature optimum (65 °C), and the pH optimum of NfPG III was the lowest (3.5). The orders of their thermostability and resistance to chemicals tested were NfPG II>NfPG III>NfPG I and NfPG II>NfPG I>NfPG III, respectively. Combinations of these enzymes showed better performance than individuals in the processing and clarification of apple and strawberry juice. These results suggest that N. fischeri polygalacturonases have great application potentials in the food industry for juice production.
Sigma-Aldrich
Polygalacturonic acid sodium salt, from citrus fruit, ≥75% (titration)
ACE microscale glassware kit, MICRO/MINI-Lab® Basic kit II
ACE microscale glassware kit, MICRO/MINI-Lab® Deluxe kit