MilliporeSigma
  • Rational Topological Design for Fluorescence Enhancement upon Aggregation of Distyrylfuran Derivatives.
Rational Topological Design for Fluorescence Enhancement upon Aggregation of Distyrylfuran Derivatives.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-04-14)
Charlotte Mallet, Chady Moussallem, Alexandre Faurie, Magali Allain, Frédéric Gohier, William G Skene, Pierre Frère
ABSTRACT
A series of 2,5-distyrylfuran derivatives bearing pentafluorophenyl- and cyanovinyl units have been synthesized for aggregation-induced emission (AIE). The effect of the type and extent of the supramolecular connections on the AIE of the furan derivatives were examined and correlated with their X-ray crystal structures. It was found that the simultaneous presence of cyano and perfluorophenyl units strongly enhances the fluorescence upon aggregation. Single-crystal X-ray diffraction analysis confirmed that CH⋅⋅⋅F, F⋅⋅⋅F, CH⋅⋅⋅nitrile, Ar⋅⋅⋅ArF (Ar=aryl, ArF =fluoroaryl), and nitrile⋅⋅⋅ArF intra- and intermolecular interactions drive the topology of the molecule and that solid-state supramolecular contacts favor AIE of the furan derivatives.
Sigma-Aldrich
3-Nitrobenzyl alcohol, 98%