MilliporeSigma
  • Antiproliferative and Pro-Apoptotic Activity of Diarylheptanoids Isolated from the Bark of Alnus japonica in Human Leukemia Cell Lines.
Antiproliferative and Pro-Apoptotic Activity of Diarylheptanoids Isolated from the Bark of Alnus japonica in Human Leukemia Cell Lines.
The American journal of Chinese medicine (2015-06-30)
Takuhiro Uto, Nguyen Huu Tung, Regina Appiah-Opong, Abigail Aning, Osamu Morinaga, Dominic Edoh, Alexander K Nyarko, Yukihiro Shoyama
ABSTRACT
Alnus japonica Steud is a tree that grows in damp areas of mountain valleys and has been used as a traditional medicine in Asia. We investigated the antiproliferative activity of hirsutanone (Hir) and oregonin (Ore) in human cancer cell lines and elucidated their mechanisms of action. A cytotoxicity study using a panel of 12 human cancer and 4 normal cell lines indicated that Hir exhibited potent antiproliferative activity against 4 leukemia (Jurkat, U937, THP-1, and HL-60) and 2 colon cancer cell lines (HCT-15 and Colo205). Although Ore suppressed the cell growth of Jurkat and THP-1, its inhibitory potency was weaker than that of Hir. The IC50 values of Hir and Ore in Jurkat were 11.37 μM and 22.16 μM, respectively. Further analysis on Jurkat cells demonstrated that Hir caused a sequence of events involved in apoptosis, including nuclear morphological changes and accumulation of cells with sub-G1 DNA content. Hir led to the cleavage of poly(ADP-ribose) polymerase (PARP) and activation of caspase-3, -8, and -9. In addition, Hir-induced PARP cleavage was completely abolished by specific inhibitors to these caspases. Our data suggested that Hir is a potent antiproliferative compound against the 4 leukemia cell lines and the 2 colon cancer cell lines tested. Furthermore, Hir exerts antiproliferative actions via caspase-dependent apoptotic cell death.
Product Description
Chang Liver (HeLa derivative) cell line
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)