MilliporeSigma
  • Catecholamine polymers as surface modifiers for enhancing interfacial strength of fiber-reinforced composites.
Catecholamine polymers as surface modifiers for enhancing interfacial strength of fiber-reinforced composites.
Composites Science and Technology (2015)
Lee W, et al.