MilliporeSigma
  • Using high-performance quantitative NMR (HP-qNMR?) for certifying traceable and highly accurate purity values of organic reference materials with uncertainties< 0.1%.
Using high-performance quantitative NMR (HP-qNMR?) for certifying traceable and highly accurate purity values of organic reference materials with uncertainties&lt; 0.1%.
Accreditation and Quality Assurance (2013)
Weber M, et al.