- Structure of the human Mdmx protein bound to the p53 tumor suppressor transactivation domain.
Structure of the human Mdmx protein bound to the p53 tumor suppressor transactivation domain.
The Mdmx oncoprotein has only recently emerged as a critical-independent to Mdm2-regulator of p53 activation. We have determined the crystal structure of the N-terminal domain of human Mdmx bound to a 15-residue transactivation domain peptide of human p53. The structure shows why antagonists of the Mdm2 binding to p53 are ineffective in the Mdmx-p53 interaction.