MilliporeSigma
  • Male germ cell expression of the PAS domain kinase PASKIN and its novel target eukaryotic translation elongation factor eEF1A1.
Male germ cell expression of the PAS domain kinase PASKIN and its novel target eukaryotic translation elongation factor eEF1A1.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2007-06-28)
Katrin Eckhardt, Juliane Troger, Jana Reissmann, Dörthe M Katschinski, Klaus F Wagner, Petra Stengel, Uwe Paasch, Peter Hunziker, Emanuela Borter, Sandra Barth, Philipp Schlafli, Patrick Spielmann, Daniel P Stiehl, Gieri Camenisch, Roland H Wenger
ABSTRACT
PASKIN links energy flux and protein synthesis in yeast, regulates glycogen synthesis in mammals, and has been implicated in glucose-stimulated insulin production in pancreatic beta-cells. Using newly generated monoclonal antibodies, PASKIN was localized in the nuclei of human testis germ cells and in the midpiece of human sperm tails. A speckle-like nuclear pattern was observed for endogenous PASKIN in HeLa cells in addition to its cytoplasmic localization. By yeast two-hybrid screening, we identified the multifunctional eukaryotic translation elongation factor eEF1A1 as a novel interaction partner of PASKIN. This interaction was mapped to the PAS A and kinase domains of PASKIN and to the C-terminus of eEF1A1 using mammalian two-hybrid and GST pull-down assays. Kinase assays, mass spectrometry and site-directed mutagenesis revealed PASKIN auto-phosphorylation as well as eEF1A1 target phosphorylation mainly but not exclusively at Thr432. Wild-type but not kinase-inactive PASKIN increased the in vitro translation of a reporter cRNA. Whereas eEF1A1 did not localize to the nucleus, it co-localizes with PASKIN to the cytoplasm of HeLa cells. The two proteins also showed a remarkably similar localization in the midpiece of the sperm tail. These data suggest regulation of eEF1A1 by PASKIN-dependent phosphorylation in somatic as well as in sperm cells.