MilliporeSigma
  • Clinicopathology of spinocerebellar degeneration: its correlation to the unstable CAG repeat of the affected gene.
Clinicopathology of spinocerebellar degeneration: its correlation to the unstable CAG repeat of the affected gene.
Pathology international (1997-01-01)
S Yagishita, M Inoue
ABSTRACT
The recent advances in gene analysis have greatly facilitated the classification of autosomal dominant spinocerebellar ataxia (SCA). Analyses of linkage in large families with SCA have assigned gene foci to at least 8 chromosomes. One gene is located in the short arm of chromosome 6 (6p22-p23) and causes spinocerebellar ataxia type 1 (SCA1). A gene in the long arm of chromosome 14 (14q24.3-q32) underlies Machado-Joseph disease (MJD). A third gene locus is assigned to the short arm of chromosome 12 (12p2-pter) causing dentatorubropallidoluysian atrophy (DRPLA). The gene for spinocerebellar ataxia type 2 (SCA2) is located in the 12q23-24. Subsequently, a sporadic counterpart of hereditary olivopontocerebellar atrophy of the Menzel type is clearly defined, and all the syndromes (non-hereditary olivopontocerebellar atrophy, striatonigral degeneration and Shy-Drager syndrome) are now lumped under the term of multiple system atrophy (MSA). Oligodendroglial cytoplasmic inclusions appear to be specific for and diagnostic of MSA. As the clinical features in SCA are variable and often appear to overlap with one another, which makes accurate classification difficult if not possible, the genotype is required for their unequivocal classification. However, major neuropathological features clearly distinguish SCA1 from SCA3/ MJD cases; the medial segment of the globus pallidus and intermediolateral column lesions in SCA3/MJD, and inferior olive and cerebellar cortical degeneration in SCA1. It has been stated that neurodegeneration in SCA3/MJD is more homogeneous than in SCA1 or SCA2 and that degeneration of the pallidoluysian system is not present in the latter. The pertinent pathology in each of the three types of SCA is illustrated. The background of clinicopathology and genetic analysis of dentatorubropallidoluysian atrophy is also reviewed.