MilliporeSigma
  • Enaptin, a giant actin-binding protein, is an element of the nuclear membrane and the actin cytoskeleton.
Enaptin, a giant actin-binding protein, is an element of the nuclear membrane and the actin cytoskeleton.
Experimental cell research (2004-04-20)
V C Padmakumar, Sabu Abraham, Stephan Braune, Angelika A Noegel, Budi Tunggal, Iakowos Karakesisoglou, Elena Korenbaum
ABSTRACT
Enaptin belongs to a family of recently identified giant proteins that associate with the F-actin cytoskeleton as well as the nuclear membrane. It is composed of an N-terminal alpha-actinin type actin-binding domain (ABD) followed by a long coiled coil rod and a transmembrane domain at the C-terminus. The ABD binds to F-actin in vivo and in vitro and leads to bundle formation. The human Enaptin gene spreads over 515 kb and gives rise to several splicing isoforms (Nesprin-1, Myne-1, Syne-1, CPG2). The longest assembled cDNA encompasses 27,669 bp and predicts a 1014 kDa protein. Antibodies against the ABD of Enaptin localise the protein at F-actin-rich structures throughout the cell and in focal contacts as well as at the nuclear envelope. In COS7 cells, the protein is also present within the nuclear compartment. With the discovery of the actin-binding properties of Enaptin and the highly homologous Nuance, we define a family of proteins that integrate the cytoskeleton with the nucleoskeleton.