MilliporeSigma
  • Microcephalin is a DNA damage response protein involved in regulation of CHK1 and BRCA1.
Microcephalin is a DNA damage response protein involved in regulation of CHK1 and BRCA1.
The Journal of biological chemistry (2004-06-29)
Xingzhi Xu, Juhie Lee, David F Stern
ABSTRACT
Microcephalin (MCPH1) is the first gene identified among at least six loci that contribute to the autosomal recessive disease, primary microcephaly. MCPH1, like NFBD1/MDC1, 53BP1, and BRCA1, encodes a protein with twin carboxyl-terminal BRCT domains (PTCB). Here, we report that Mcph1 forms ionizing radiation-induced foci. Down-regulation of Mcph1, like other PTCBs, by siRNA, impairs ionizing radiation-induced intra-S-phase and G(2)/M checkpoints. Inhibition of the expression of Mcph1 decreases both protein and transcript levels of endogenous Brca1 but not exogenous Brca1. Mcph1 inhibition also decreases both endogenous and heterologous Chk1 transcripts and protein. We conclude that Mcph1 is involved in DNA damage-induced cellular responses, and we propose that regulation of Brca1 and/or Chk1 by Mcph1 may contribute to these cellular responses.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution